Product Details

SNP ID
rs79005297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:83956884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGTCACTTATTCTCTGAAGCAC[A/C]CCATGATCTCTCTCCACAAGTTAAT
Phenotype
MIM: 603961
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SEMA3A PubMed Links

Gene Details

Gene
SEMA3A
Gene Name
semaphorin 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006080.2 7153 Intron NP_006071.1
XM_005250110.3 7153 UTR 3 XP_005250167.1
XM_005250111.4 7153 UTR 3 XP_005250168.1
XM_006715839.3 7153 UTR 3 XP_006715902.1
XM_011515734.2 7153 UTR 3 XP_011514036.1
XM_017011673.1 7153 UTR 3 XP_016867162.1

View Full Product Details