Product Details

SNP ID
rs79481528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93426070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTATCTGAACTAGGTCAATTTC[A/C]TTGTTTTTCTTTGATACTTTGCTTG
Phenotype
MIM: 114131
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CALCR PubMed Links

Gene Details

Gene
CALCR
Gene Name
calcitonin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164737.1 1994 UTR 3 NP_001158209.1
NM_001164738.1 1994 UTR 3 NP_001158210.1
NM_001742.3 1994 UTR 3 NP_001733.1

View Full Product Details