Product Details
- SNP ID
-
rs79217451
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:132711454 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCGTAGAACAGCCTGTAGGGTAA[C/T]AGGTGGATTTTCCCCATATTTTTAT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM71
PubMed Links
Gene Details
- Gene
- TMEM71
- Gene Name
- transmembrane protein 71
View Full Product Details