Product Details

SNP ID
rs78805444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18529965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCAAAGACTATTATTCATCTTC[A/G]AATAGTCAAATTCTAACACAGTATT
Phenotype
MIM: 601387 MIM: 610985
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSG101 PubMed Links

Gene Details

Gene
TSG101
Gene Name
tumor susceptibility 101
There are no transcripts associated with this gene.

Gene
UEVLD
Gene Name
UEV and lactate/malate dehyrogenase domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040697.2 5694 Intron NP_001035787.1
NM_001261382.1 5694 Intron NP_001248311.1
NM_001261383.1 5694 Intron NP_001248312.1
NM_001261384.1 5694 Intron NP_001248313.1
NM_001261385.1 5694 Intron NP_001248314.1
NM_001261386.1 5694 Intron NP_001248315.1
NM_001297771.1 5694 Intron NP_001284700.1
NM_018314.4 5694 Intron NP_060784.3
XM_006718255.2 5694 UTR 3 XP_006718318.1
XM_011520201.2 5694 UTR 3 XP_011518503.1
XM_017017985.1 5694 UTR 3 XP_016873474.1
XM_017017986.1 5694 UTR 3 XP_016873475.1

View Full Product Details