Product Details

SNP ID
rs75188962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8920120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAGAGGTGAAACAAAATGTCACT[C/T]ATGTACAACTAGGCTTTATGGGAAG
Phenotype
MIM: 609191 MIM: 140750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKIP1 PubMed Links

Gene Details

Gene
AKIP1
Gene Name
A-kinase interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206646.1 2342 Intron NP_001193575.1
NM_001206647.1 2342 Intron NP_001193576.1
NM_001206648.1 2342 Intron NP_001193577.1
NM_020642.3 2342 Intron NP_065693.2
XM_017018011.1 2342 Intron XP_016873500.1
XM_017018012.1 2342 Intron XP_016873501.1
Gene
C11orf16
Gene Name
chromosome 11 open reading frame 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020643.2 2342 UTR 3 NP_065694.2
XM_017018013.1 2342 UTR 3 XP_016873502.1
Gene
ST5
Gene Name
suppression of tumorigenicity 5
There are no transcripts associated with this gene.

View Full Product Details