Product Details

SNP ID
rs75768734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1619954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGACAATGCGAGGTCAGGAGG[A/G]GTCCCGGTTTGCTCACCGACTGCCT
Phenotype
MIM: 614620
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRAMP1 PubMed Links

Gene Details

Gene
CRAMP1
Gene Name
cramped chromatin regulator homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020825.3 Intron NP_065876.3
Gene
IFT140
Gene Name
intraflagellar transport 140
There are no transcripts associated with this gene.

Gene
LOC105371046
Gene Name
uncharacterized LOC105371046
There are no transcripts associated with this gene.

View Full Product Details