Product Details

SNP ID
rs74345637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:22100303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACCTTGGCTGACCTGATAAGCC[A/T]GGGCACAGAAGTGCTGTAAGTCTGA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
VWA3A PubMed Links

Gene Details

Gene
VWA3A
Gene Name
von Willebrand factor A domain containing 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173615.4 601 Missense Mutation CAG,CTG Q112L NP_775886.3
XM_011545742.2 601 Missense Mutation CAG,CTG Q112L XP_011544044.1
XM_011545744.2 601 Intron XP_011544046.1
XM_011545745.1 601 Intron XP_011544047.1
XM_011545746.2 601 Intron XP_011544048.1
XM_017022947.1 601 Missense Mutation CAG,CTG Q112L XP_016878436.1
XM_017022948.1 601 Intron XP_016878437.1
XM_017022949.1 601 Intron XP_016878438.1

View Full Product Details