Product Details

SNP ID
rs76274703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75649651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCCACAGACTCCTTTTTAAAAA[A/G]GATTGCTGTTATGTTCAGGTAGATT
Phenotype
MIM: 601421 MIM: 605061
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KARS PubMed Links

Gene Details

Gene
KARS
Gene Name
lysyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
TERF2IP
Gene Name
TERF2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018975.3 Intron NP_061848.2

View Full Product Details