Product Details

SNP ID
rs78027392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49430551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGGACAGGGGGTGGGGGCCTGG[G/A]GGGCTGAAATGTGCTGTGTGTGGCC
Phenotype
MIM: 608386 MIM: 605208
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
GFY PubMed Links

Gene Details

Gene
GFY
Gene Name
golgi associated olfactory signaling regulator
There are no transcripts associated with this gene.

Gene
PTH2
Gene Name
parathyroid hormone 2
There are no transcripts associated with this gene.

Gene
SLC17A7
Gene Name
solute carrier family 17 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020309.3 1823 Missense Mutation CCC,TCC P551S NP_064705.1

View Full Product Details