Product Details

SNP ID
rs80239416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36892281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAATTCTCTGATGTTGGATAAA[C/T]TGTGAGTTTTGATTAAAGGTCTTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF345 PubMed Links

Gene Details

Gene
ZNF345
Gene Name
zinc finger protein 345
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242472.1 875 Intron NP_001229401.1
NM_001242474.1 875 Intron NP_001229403.1
NM_001242475.1 875 Intron NP_001229404.1
NM_001242476.1 875 Intron NP_001229405.1
NM_003419.4 875 Intron NP_003410.1
XM_011526704.2 875 Intron XP_011525006.1
XM_017026568.1 875 Intron XP_016882057.1
XM_017026569.1 875 Intron XP_016882058.1
XM_017026570.1 875 Intron XP_016882059.1
XM_017026571.1 875 Intron XP_016882060.1
XM_017026572.1 875 Intron XP_016882061.1
XM_017026573.1 875 Intron XP_016882062.1
Gene
ZNF829
Gene Name
zinc finger protein 829
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037232.3 875 Silent Mutation CAA,CAG Q170Q NP_001032309.2
NM_001171979.1 875 Silent Mutation CAA,CAG Q251Q NP_001165450.1
XM_005258876.4 875 Silent Mutation CAA,CAG Q170Q XP_005258933.1
XM_011526933.2 875 Silent Mutation CAA,CAG Q170Q XP_011525235.1

View Full Product Details