Product Details
- SNP ID
-
rs77693690
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:19761208 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATTGCATTTTGTAGGTGAGGAAA[C/T]GAAAGCCTAAAATTAGTCTGTCATT
- Phenotype
-
MIM: 610222
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC107987276
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143422567] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC107987276
- Gene Name
- translation initiation factor IF-2-like
- Gene
- RIN2
- Gene Name
- Ras and Rab interactor 2
View Full Product Details