Product Details

SNP ID
rs74505338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:328717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGATGCCAGGACTGAATAAAGT[A/G]TATTTGCCCCGACCTTGCCCTGTGG
Phenotype
MIM: 601947
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NRSN2-AS1 PubMed Links

Gene Details

Gene
NRSN2-AS1
Gene Name
NRSN2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SOX12
Gene Name
SRY-box 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006943.3 3147 UTR 3 NP_008874.2

View Full Product Details