Product Details

SNP ID
rs75205692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25613452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGTTCTAGGCCCACCTCTGTAA[A/G]CAGTGTTAGGGAGGGCCTCTGGCTT
Phenotype
MIM: 610763
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NANP PubMed Links

Gene Details

Gene
NANP
Gene Name
N-acetylneuraminic acid phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152667.2 3284 UTR 3 NP_689880.1

View Full Product Details