Product Details

SNP ID
rs79395339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3755220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCCTCAGCAGCTCCCATCTTGG[C/T]TACTGCCCCACGGAGGTAACCATCA
Phenotype
MIM: 610702
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf27 PubMed Links

Gene Details

Gene
C20orf27
Gene Name
chromosome 20 open reading frame 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039140.2 Intron NP_001034229.1
NM_001258429.1 Intron NP_001245358.1
NM_001258430.1 Intron NP_001245359.1
XM_011529266.2 Intron XP_011527568.1
Gene
HSPA12B
Gene Name
heat shock protein family A (Hsp70) member 12B
There are no transcripts associated with this gene.

View Full Product Details