Product Details

SNP ID
rs77910593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33071800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGAGAGCTGTATTTAAGACTCG[C/T]GTATCCAGTGTTTTGTCGCAGAGAG
Phenotype
MIM: 606385
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00945 PubMed Links

Gene Details

Gene
LINC00945
Gene Name
long intergenic non-protein coding RNA 945
There are no transcripts associated with this gene.

Gene
OLIG1
Gene Name
oligodendrocyte transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138983.2 1657 UTR 3 NP_620450.2

View Full Product Details