Product Details
- SNP ID
-
rs77409148
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:56565318 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGAGCTTCACTCCAGGTGTGGGGG[G/T]GGGGGTGCTGGGAACCGCTATAGGC
- Phenotype
-
MIM: 300264
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
UBQLN2
PubMed Links
Gene Details
- Gene
- UBQLN2
- Gene Name
- ubiquilin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013444.3 |
1726 |
Missense Mutation |
GGG,GTG |
G482V |
NP_038472.2 |
View Full Product Details