Product Details

SNP ID
rs17852080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:112145696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGGTGGGATCCCGGGGCGAGCC[A/C]GCGGCGGCGCGGACAGCGGCCGAAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C4orf32 PubMed Links

Gene Details

Gene
C4orf32
Gene Name
chromosome 4 open reading frame 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152400.2 300 Missense Mutation CAG,CCG Q39P NP_689613.1
XM_017007732.1 300 UTR 5 XP_016863221.1

View Full Product Details