Product Details

SNP ID
rs75872488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:165040215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTGTATCCTGGAAGGATCTAG[A/T]TGATACCTTTCCCTGTCCTGTCTGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TRIM60 PubMed Links

Gene Details

Gene
TRIM60
Gene Name
tripartite motif containing 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258025.1 307 Missense Mutation GAT,GTT D48V NP_001244954.1
NM_152620.2 307 Missense Mutation GAT,GTT D48V NP_689833.1
XM_011531683.2 307 Missense Mutation GAT,GTT D48V XP_011529985.1

View Full Product Details