Product Details

SNP ID
rs76140495
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:8993935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCACAGCGCCTGGCCACCCAGAG[A/G]ATGTTCTCTTTATTCAAGGGTACAA
Phenotype
MIM: 610371
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC2A7 PubMed Links

Gene Details

Gene
SLC2A7
Gene Name
solute carrier family 2 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207420.2 Intron NP_997303.2
XM_011540824.2 Intron XP_011539126.1
XM_011540825.2 Intron XP_011539127.1

View Full Product Details