Product Details

SNP ID
rs79553963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85145546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTCAGGGTCATTAAACTCAGCC[A/G]GATTTGCAAGACATACCTTCTAGAG
Phenotype
MIM: 604652
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCF7L1 PubMed Links

Gene Details

Gene
TCF7L1
Gene Name
transcription factor 7 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031283.2 Intron NP_112573.1
XM_006712109.2 Intron XP_006712172.1

View Full Product Details