Product Details

SNP ID
rs75929826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:112311372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCATCAAAAATATGCATCTCCAT[A/G]AAAATAAATCAGCATTCAAATGAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927640 PubMed Links

Gene Details

Gene
LOC101927640
Gene Name
uncharacterized LOC101927640
There are no transcripts associated with this gene.

Gene
RFPL4B
Gene Name
ret finger protein like 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013734.2 Intron NP_001013756.2
XM_017010897.1 Intron XP_016866386.1
XM_017010898.1 Intron XP_016866387.1

View Full Product Details