Product Details

SNP ID
rs75158054
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:31483620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCGATGGCCGAAAGGAAGAAAA[C/T]AGAGGTGAACATCCCCAGAGACAAA
Phenotype
MIM: 610118
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GPR33 PubMed Links

Gene Details

Gene
GPR33
Gene Name
G protein-coupled receptor 33 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197184.2 401 Missense Mutation ATT,GTT I116V NP_001184113.2

View Full Product Details