Product Details
- SNP ID
-
rs78982717
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
24
- Location
-
Chr.1:234759347 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGGAGCCCCAAAAGATATTGGGA[C/G]CAAAACAAATGTATCTCAATGGCAT
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PP2672
PubMed Links
Gene Details
- Gene
- PP2672
- Gene Name
- uncharacterized LOC100130249
There are no transcripts associated with this gene.
View Full Product Details