Product Details

SNP ID
rs80068230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:17494647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGATTCTATTTTCTCTTGTCTCAT[A/T]GTCTGTCTTAAGAGACTCATACTAA
Phenotype
MIM: 601872
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC7A2 PubMed Links

Gene Details

Gene
SLC7A2
Gene Name
solute carrier family 7 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008539.3 Intron NP_001008539.3
NM_001164771.1 Intron NP_001158243.1
NM_003046.5 Intron NP_003037.4
XM_005273610.4 Intron XP_005273667.1
XM_005273611.4 Intron XP_005273668.1
XM_005273612.4 Intron XP_005273669.1
XM_017013746.1 Intron XP_016869235.1
XM_017013747.1 Intron XP_016869236.1

View Full Product Details