Product Details

SNP ID
rs78181205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68044255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACCTGGCCCTGAACCAGTGCAGC[A/G]TGAGCACCACGCACAAGTGCCTCAT
Phenotype
MIM: 118491 MIM: 602141 MIM: 604592
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHKA PubMed Links

Gene Details

Gene
CHKA
Gene Name
choline kinase alpha
There are no transcripts associated with this gene.

Gene
MIR6753
Gene Name
microRNA 6753
There are no transcripts associated with this gene.

Gene
NDUFS8
Gene Name
NADH:ubiquinone oxidoreductase core subunit S8
There are no transcripts associated with this gene.

Gene
TCIRG1
Gene Name
T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006019.3 1055 Missense Mutation ATG,GTG M311V NP_006010.2
NM_006053.3 1055 Missense Mutation ATG,GTG M95V NP_006044.1
XM_005273709.3 1055 Missense Mutation ATG,GTG M311V XP_005273766.1
XM_011544726.2 1055 Missense Mutation ATG,GTG M311V XP_011543028.1
XM_017017089.1 1055 Missense Mutation ATG,GTG M311V XP_016872578.1
XM_017017090.1 1055 Missense Mutation ATG,GTG M13V XP_016872579.1

View Full Product Details