Product Details

SNP ID
rs76285432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58528197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTCCTTCTCATGAAAGCCTGGA[A/G]AGATAAAATCAGGAATTCACTGTTG
Phenotype
MIM: 605390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LPXN PubMed Links

Gene Details

Gene
LPXN
Gene Name
leupaxin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143995.2 Intron NP_001137467.1
NM_001307951.1 Intron NP_001294880.1
NM_004811.2 Intron NP_004802.1
XM_006718750.1 Intron XP_006718813.1
XM_011545388.2 Intron XP_011543690.1
XM_011545390.2 Intron XP_011543692.1
XM_011545391.2 Intron XP_011543693.1
XM_011545393.2 Intron XP_011543695.1
XM_011545394.2 Intron XP_011543696.1
XM_017018579.1 Intron XP_016874068.1

View Full Product Details