Product Details

SNP ID
rs79855615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60928194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAAGGATAGCTTACGGCAGAGT[A/C]ATCAATCAGCCAGTGTGTTTGCATT
Phenotype
MIM: 610408
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC15A3 PubMed Links

Gene Details

Gene
SLC15A3
Gene Name
solute carrier family 15 member 3
There are no transcripts associated with this gene.

Gene
TMEM109
Gene Name
transmembrane protein 109
There are no transcripts associated with this gene.

Gene
TMEM132A
Gene Name
transmembrane protein 132A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017870.3 Intron NP_060340.2
NM_178031.2 Intron NP_821174.1
XM_017017951.1 Intron XP_016873440.1
XM_017017952.1 Intron XP_016873441.1

View Full Product Details