Product Details

SNP ID
rs114859769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:8858540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCACTCCATTTCCAATCTGGTG[A/T]ACAGGAAATTCCTTATCTATAACTG
Phenotype
MIM: 611949
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MBOAT2 PubMed Links

Gene Details

Gene
MBOAT2
Gene Name
membrane bound O-acyltransferase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321265.1 1915 UTR 3 NP_001308194.1
NM_001321266.1 1915 UTR 3 NP_001308195.1
NM_001321267.1 1915 UTR 3 NP_001308196.1
NM_138799.3 1915 UTR 3 NP_620154.2
XM_011510313.1 1915 Intron XP_011508615.1
XM_011510314.2 1915 Intron XP_011508616.1

View Full Product Details