Product Details

SNP ID
rs114025250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160436272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTTTATGTGCAATCTGTAGGGC[C/G]AGGAGCATAGTCTAATCTGACCCAG
Phenotype
MIM: 605575 MIM: 616148
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SMC4 PubMed Links

Gene Details

Gene
SMC4
Gene Name
structural maintenance of chromosomes 4
There are no transcripts associated with this gene.

Gene
TRIM59
Gene Name
tripartite motif containing 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173084.2 3107 UTR 3 NP_775107.1

View Full Product Details