Product Details

SNP ID
rs116752753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101574253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCGGGAGACGAGAAGCCTGAAA[A/G]GTCGCAGCGAGCTGGAGCCGCCGGA
Phenotype
MIM: 615210 MIM: 615423
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCNP PubMed Links

Gene Details

Gene
PCNP
Gene Name
PEST proteolytic signal containing nuclear protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320395.1 160 Intron NP_001307324.1
NM_001320397.1 160 Intron NP_001307326.1
NM_001320398.1 160 Intron NP_001307327.1
NM_001320399.1 160 UTR 5 NP_001307328.1
NM_001320400.1 160 Intron NP_001307329.1
NM_001320401.1 160 Intron NP_001307330.1
NM_020357.2 160 Intron NP_065090.1
XM_017006922.1 160 Intron XP_016862411.1
Gene
TRMT10C
Gene Name
tRNA methyltransferase 10C, mitochondrial RNase P subunit
There are no transcripts associated with this gene.

View Full Product Details