Product Details

SNP ID
rs115287053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34424876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAACTACTGTTAAGGCGTTAAGT[A/C]GAAGCTTGGATCATCCTAGGCAAGT
Phenotype
MIM: 603632
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RPS10 PubMed Links

Gene Details

Gene
RPS10
Gene Name
ribosomal protein S10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014.4 Intron NP_001005.1
NM_001203245.2 Intron NP_001190174.1
NM_001204091.1 Intron NP_001191020.1
Gene
RPS10-NUDT3
Gene Name
RPS10-NUDT3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202470.2 Intron NP_001189399.1

View Full Product Details