Product Details

SNP ID
rs116685719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151379270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGGGGAGGTTGGATGCCATTAGA[A/G]GAGCAGGAGTGGGCATGGGCGTGTA
Phenotype
MIM: 607981
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUB1 PubMed Links

Gene Details

Gene
NUB1
Gene Name
negative regulator of ubiquitin like proteins 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243351.1 2727 Intron NP_001230280.1
NM_016118.4 2727 Intron NP_057202.3
XM_005250011.2 2727 Intron XP_005250068.1
XM_017012304.1 2727 Intron XP_016867793.1
XM_017012305.1 2727 Intron XP_016867794.1
XM_017012306.1 2727 Intron XP_016867795.1
XM_017012307.1 2727 Intron XP_016867796.1
XM_017012308.1 2727 Intron XP_016867797.1
XM_017012309.1 2727 Intron XP_016867798.1
XM_017012310.1 2727 Intron XP_016867799.1
XM_017012311.1 2727 Intron XP_016867800.1
XM_017012312.1 2727 Intron XP_016867801.1
XM_017012313.1 2727 Intron XP_016867802.1
XM_017012314.1 2727 Intron XP_016867803.1
Gene
WDR86
Gene Name
WD repeat domain 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284260.1 2727 Intron NP_001271189.1
NM_001284261.1 2727 Intron NP_001271190.1
NM_001284262.1 2727 Intron NP_001271191.1
NM_198285.2 2727 Intron NP_938026.2
XM_005249989.3 2727 Intron XP_005250046.1
XM_005249990.4 2727 Intron XP_005250047.1
XM_006715966.2 2727 Intron XP_006716029.1
XM_011516144.2 2727 UTR 3 XP_011514446.1
XM_011516145.2 2727 UTR 3 XP_011514447.1
XM_011516146.2 2727 Intron XP_011514448.1
XM_011516147.2 2727 UTR 3 XP_011514449.1
XM_011516148.1 2727 Intron XP_011514450.1
XM_011516150.1 2727 Intron XP_011514452.1
XM_011516151.2 2727 UTR 3 XP_011514453.1
XM_011516152.1 2727 Intron XP_011514454.1

View Full Product Details