Product Details

SNP ID
rs114175550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33244324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCTGCCTTAACAAACTGCATTG[C/T]GTCACCTCTTTGAGCCTCTTTCGTA
Phenotype
MIM: 601497 MIM: 613929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAG1 PubMed Links

Gene Details

Gene
BAG1
Gene Name
BCL2 associated athanogene 1
There are no transcripts associated with this gene.

Gene
SPINK4
Gene Name
serine peptidase inhibitor, Kazal type 4
There are no transcripts associated with this gene.

View Full Product Details