Product Details
- SNP ID
-
rs115073289
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:127742048 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCGCTGCCTGTGGTTGGGCCGGGA[A/G]AGGCGGAGGGCGGAGCCAACGTGAG
- Phenotype
-
MIM: 604722
MIM: 608052
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SH2D3C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3217145] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SH2D3C
- Gene Name
- SH2 domain containing 3C
- Gene
- TOR2A
- Gene Name
- torsin family 2 member A
There are no transcripts associated with this gene.
View Full Product Details