Product Details

SNP ID
rs117802349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3799022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATACATTTGCCAAGCTTTCACAG[C/T]GAGTTGAGCTAATGCTTGGGCAGGC
Phenotype
MIM: 601021 MIM: 615187
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUP98 PubMed Links

Gene Details

Gene
NUP98
Gene Name
nucleoporin 98
There are no transcripts associated with this gene.

Gene
PGAP2
Gene Name
post-GPI attachment to proteins 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145438.2 Intron NP_001138910.1
NM_001256235.1 Intron NP_001243164.1
NM_001256236.1 Intron NP_001243165.1
NM_001256237.1 Intron NP_001243166.1
NM_001256238.1 Intron NP_001243167.1
NM_001256239.1 Intron NP_001243168.1
NM_001256240.1 Intron NP_001243169.1
NM_001283038.1 Intron NP_001269967.1
NM_001283039.1 Intron NP_001269968.1
NM_001283040.1 Intron NP_001269969.1
NM_014489.3 Intron NP_055304.1
XM_006718181.3 Intron XP_006718244.1
XM_006718183.3 Intron XP_006718246.1
XM_006718185.2 Intron XP_006718248.1
XM_006718186.2 Intron XP_006718249.1
XM_006718190.3 Intron XP_006718253.1
XM_006718191.3 Intron XP_006718254.1
XM_006718192.1 Intron XP_006718255.1
XM_006718193.2 Intron XP_006718256.1
XM_011519990.2 Intron XP_011518292.2
XM_011519991.2 Intron XP_011518293.2
XM_011519992.1 Intron XP_011518294.1
XM_011519993.2 Intron XP_011518295.2
XM_011519994.2 Intron XP_011518296.2
XM_011519995.2 Intron XP_011518297.1
XM_011519996.1 Intron XP_011518298.1
XM_011519997.1 Intron XP_011518299.1
XM_011519998.2 Intron XP_011518300.1
XM_011519999.1 Intron XP_011518301.1
XM_011520000.1 Intron XP_011518302.1
XM_011520001.1 Intron XP_011518303.1
XM_011520002.1 Intron XP_011518304.1
XM_011520003.2 Intron XP_011518305.1
XM_011520004.2 Intron XP_011518306.1
XM_011520005.2 Intron XP_011518307.1
XM_011520006.2 Intron XP_011518308.1
XM_011520007.2 Intron XP_011518309.1
XM_017017556.1 Intron XP_016873045.1
XM_017017557.1 Intron XP_016873046.1
XM_017017558.1 Intron XP_016873047.1
XM_017017559.1 Intron XP_016873048.1
XM_017017560.1 Intron XP_016873049.1

View Full Product Details