Product Details

SNP ID
rs118062148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:91404307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTATAGATGAATGGTCTCTTGGT[A/G]TAACTGTTCAACCGACAAACAACAG
Phenotype
MIM: 602446
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPC5 PubMed Links

Gene Details

Gene
GPC5
Gene Name
glypican 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004466.5 Intron NP_004457.1
XM_011521054.2 Intron XP_011519356.1
XM_011521055.2 Intron XP_011519357.1
XM_011521056.2 Intron XP_011519358.1
XM_011521057.2 Intron XP_011519359.1
XM_011521058.2 Intron XP_011519360.1
XM_011521059.2 Intron XP_011519361.1
XM_011521060.2 Intron XP_011519362.1
XM_017020435.1 Intron XP_016875924.1
XM_017020436.1 Intron XP_016875925.1
XM_017020437.1 Intron XP_016875926.1

View Full Product Details