Product Details

SNP ID
rs117683177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48966882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAAACATTTATTGGGAAGGGAG[A/G]TTCTGTCATCTTCGGACAATTAACC
Phenotype
MIM: 137240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GIP PubMed Links

Gene Details

Gene
GIP
Gene Name
gastric inhibitory polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004123.2 Intron NP_004114.1

View Full Product Details