Product Details

SNP ID
rs116222735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44027462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGAAAAGTTTTGGGTGATGGG[A/G]ACAACAAGCCAAAGAGAAGGGAATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF222 PubMed Links

Gene Details

Gene
ZNF222
Gene Name
zinc finger protein 222
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129996.1 347 Silent Mutation GGA,GGG G78G NP_001123468.1
NM_013360.2 347 Intron NP_037492.2
XM_005259208.3 347 Silent Mutation GGA,GGG G69G XP_005259265.1
XM_005259209.3 347 Intron XP_005259266.1
XM_017027237.1 347 Silent Mutation GGA,GGG G78G XP_016882726.1
XM_017027238.1 347 Silent Mutation GGA,GGG G69G XP_016882727.1

View Full Product Details