Product Details
- SNP ID
-
rs117713780
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:55432762 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACAAGGCGTCGTCTCTGTTATTA[C/T]ACCACAAGAAGGAAGCCTCTGAGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SHISA7
PubMed Links
Gene Details
- Gene
- SHISA7
- Gene Name
- shisa family member 7
View Full Product Details