Product Details

SNP ID
rs116993790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49280370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCCGACAGATCGACTATGTTG[A/C]TCTAACTTTTCTAAGCCAGTTTCTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SNORD12 PubMed Links

Gene Details

Gene
SNORD12
Gene Name
small nucleolar RNA, C/D box 12
There are no transcripts associated with this gene.

Gene
SNORD12B
Gene Name
small nucleolar RNA, C/D box 12B
There are no transcripts associated with this gene.

Gene
SNORD12C
Gene Name
small nucleolar RNA, C/D box 12C
There are no transcripts associated with this gene.

Gene
ZFAS1
Gene Name
ZNFX1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZNFX1
Gene Name
zinc finger NFX1-type containing 1
There are no transcripts associated with this gene.

View Full Product Details