Product Details

SNP ID
rs114790387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58983954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTCTCATGGATGAGGGGGGTGG[A/G]CCAGACGTTCATTTCATGTTTATTC
Phenotype
MIM: 605297
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NELFCD PubMed Links

Gene Details

Gene
NELFCD
Gene Name
negative elongation factor complex member C/D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198976.2 Intron NP_945327.2

View Full Product Details