Product Details

SNP ID
rs114566876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20441571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTTCATTGACCAACAACATGGG[G/T]TCTGAAAACCCAGCGGGAGGGGTCT
Phenotype
MIM: 613619
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHL22 PubMed Links

Gene Details

Gene
KLHL22
Gene Name
kelch like family member 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032775.3 5124 UTR 3 NP_116164.2
XM_017029018.1 5124 UTR 3 XP_016884507.1
XM_017029019.1 5124 UTR 3 XP_016884508.1
XM_017029020.1 5124 UTR 3 XP_016884509.1
XM_017029021.1 5124 UTR 3 XP_016884510.1
XM_017029022.1 5124 UTR 3 XP_016884511.1
XM_017029023.1 5124 UTR 3 XP_016884512.1
XM_017029024.1 5124 UTR 3 XP_016884513.1
XM_017029025.1 5124 UTR 3 XP_016884514.1
Gene
SCARF2
Gene Name
scavenger receptor class F member 2
There are no transcripts associated with this gene.

View Full Product Details