Product Details

SNP ID
rs112411662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70426727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCTGTGGCTGATACCTTCTACC[A/G]TATCCACCGAAGAGGTGCCAACGCT
Phenotype
MIM: 300521
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GDPD2 PubMed Links

Gene Details

Gene
GDPD2
Gene Name
glycerophosphodiester phosphodiesterase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171191.1 419 Missense Mutation CAT,CGT H102R NP_001164662.1
NM_001171192.1 419 Missense Mutation CAT,CGT H181R NP_001164663.1
NM_001171193.1 419 Missense Mutation CAT,CGT H102R NP_001164664.1
NM_017711.3 419 Missense Mutation CAT,CGT H181R NP_060181.2
XM_011530977.1 419 Missense Mutation CAT,CGT H54R XP_011529279.1
XM_017029614.1 419 UTR 5 XP_016885103.1
Gene
KIF4A
Gene Name
kinesin family member 4A
There are no transcripts associated with this gene.

Gene
LOC105373244
Gene Name
uncharacterized LOC105373244
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011531081.2 419 Intron XP_011529383.1
XM_011531082.2 419 Intron XP_011529384.1

View Full Product Details