Product Details

SNP ID
rs114115231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:151670073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATCAAGGCCTCCAAGGATTTTA[C/T]TCTCTTACATCACAGTTTTGACAAG
Phenotype
MIM: 603645
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM160A1 PubMed Links

Gene Details

Gene
FAM160A1
Gene Name
family with sequence similarity 160 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109977.1 2643 Intron NP_001103447.1
XM_005263199.1 2643 UTR 3 XP_005263256.1
XM_011532220.1 2643 UTR 3 XP_011530522.1
XM_011532221.2 2643 UTR 3 XP_011530523.1
XM_011532222.2 2643 UTR 3 XP_011530524.1
XM_011532223.2 2643 UTR 3 XP_011530525.1
XM_011532224.2 2643 UTR 3 XP_011530526.1
XM_017008583.1 2643 UTR 3 XP_016864072.1
Gene
GATB
Gene Name
glutamyl-tRNA amidotransferase subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004564.2 2643 Intron NP_004555.1
XM_005263063.3 2643 UTR 3 XP_005263120.1

View Full Product Details