Product Details

SNP ID
rs116278971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:48451844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTTCTGTCTCTTTTTGGATAAG[C/T]CCCTGATCAGTAATATGTCTGAGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1R21 PubMed Links

Gene Details

Gene
PPP1R21
Gene Name
protein phosphatase 1 regulatory subunit 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135629.2 Intron NP_001129101.1
NM_001193475.1 Intron NP_001180404.1
NM_152994.4 Intron NP_694539.1
XM_017003301.1 Intron XP_016858790.1
XM_017003302.1 Intron XP_016858791.1

View Full Product Details