Product Details

SNP ID
rs114104592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70261105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGTTCCAAAGATAGAGCTCCTT[C/T]ATGTGTCTCCTGAGAACTGAGGGAA
Phenotype
MIM: 610995
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR1285-2 PubMed Links

Gene Details

Gene
MIR1285-2
Gene Name
microRNA 1285-2
There are no transcripts associated with this gene.

Gene
PCYOX1
Gene Name
prenylcysteine oxidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016297.3 Intron NP_057381.3

View Full Product Details