Product Details

SNP ID
rs115824995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40532112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAGAGTTGCACAGAATGCCAGT[A/G]GGAGGACTTCTCAGGAACGTTTCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF621 PubMed Links

Gene Details

Gene
ZNF621
Gene Name
zinc finger protein 621
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098414.2 628 Silent Mutation GTA,GTG V114V NP_001091884.1
NM_001287245.1 628 Silent Mutation GTA,GTG V114V NP_001274174.1
NM_198484.4 628 Silent Mutation GTA,GTG V114V NP_940886.1
XM_005265079.3 628 Silent Mutation GTA,GTG V106V XP_005265136.1
XM_005265080.3 628 Silent Mutation GTA,GTG V79V XP_005265137.1
XM_006713114.3 628 Silent Mutation GTA,GTG V106V XP_006713177.1
XM_011533610.2 628 Silent Mutation GTA,GTG V106V XP_011531912.1
XM_017006226.1 628 Silent Mutation GTA,GTG V79V XP_016861715.1

View Full Product Details