Product Details

SNP ID
rs113697253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:48411982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGAACAAAAACGGCGGGTGCAC[C/T]TCCTCCGATCACAGTTAACCACACG
Phenotype
MIM: 609239 MIM: 614041
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LPAR6 PubMed Links

Gene Details

Gene
LPAR6
Gene Name
lysophosphatidic acid receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162497.1 2067 Missense Mutation AGT,GGT S148G NP_001155969.1
NM_001162498.1 2067 Missense Mutation AGT,GGT S148G NP_001155970.1
NM_005767.5 2067 Missense Mutation AGT,GGT S148G NP_005758.2
Gene
RB1
Gene Name
RB transcriptional corepressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000321.2 2067 Intron NP_000312.2
XM_011535171.2 2067 Intron XP_011533473.1

View Full Product Details