Product Details

SNP ID
rs115819161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53210984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTGTAGGAGGGTCCCTGCCCCA[A/G]TGGCTGAGATGGAGGGCAGGAGGAG
Phenotype
MIM: 147559 MIM: 180190
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGB7 PubMed Links

Gene Details

Gene
ITGB7
Gene Name
integrin subunit beta 7
There are no transcripts associated with this gene.

Gene
RARG
Gene Name
retinoic acid receptor gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000966.5 2223 UTR 3 NP_000957.1
NM_001042728.2 2223 UTR 3 NP_001036193.1
NM_001243730.1 2223 UTR 3 NP_001230659.1
NM_001243731.1 2223 UTR 3 NP_001230660.1
NM_001243732.1 2223 UTR 3 NP_001230661.1

View Full Product Details