Product Details

SNP ID
rs117985155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55492412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACAGCAATAACAACATTCATCC[C/T]TCTGGGACTGACAGAAGATCCTCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6C68 PubMed Links

Gene Details

Gene
OR6C68
Gene Name
olfactory receptor family 6 subfamily C member 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005519.2 35 Missense Mutation CCT,CTT P12L NP_001005519.2

View Full Product Details